Gene Transcripts
- Gene Symbol
- Ifnb1
- Entrez Gene ID
- 24481
- Gene Name
- interferon beta 1, fibroblast
- Gene Aliases
- Ifnb
- Chromosome Location
- Chr. 5: 106865192 - 106865746 on Build Rnor_6.0
- UniGene ID
- Rn.138105
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019127.1 |
NP_062000.1
|
1 |
454 |
|
View Full Product Details