Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004720.5 | NP_004711.2 | 3 | 1002 | |
GenBank mRNA | AF011466.1 | - | 3 | 947 | |
AF197929.1 | - | 3 | 981 | ||
AF233092.1 | - | 2 | 869 | ||
AK123043.1 | - | 6 | 1766 | ||
AK316573.1 | - | 3 | 963 | ||
AY322548.1 | - | 2 | 863 | ||
BC025695.1 | - | 3 | 1042 | 5209267 |