Gene Transcripts

Gene Symbol
Irs3
Entrez Gene ID
84021
Gene Name
insulin receptor substrate 3
Gene Aliases
Chromosome Location
Chr. 12: 22122953 - 22125091 on Build Rnor_6.0
UniGene ID
Rn.9791
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032074.1 NP_114463.1 1 452
GenBank mRNA BC161840.1 - 1 304
U93880.1 - 1 452

View Full Product Details