Gene Transcripts

Gene Symbol
Pim3
Entrez Gene ID
64534
Gene Name
Pim-3 proto-oncogene, serine/threonine kinase
Gene Aliases
Chromosome Location
Chr. 7: 129856467 - 129863441 on Build Rnor_6.0
UniGene ID
Rn.6343
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022602.1 NP_072124.1 260
GenBank mRNA AF057026.1 - 260
AF086624.1 - 462
BC097317.1 - 239 7446368

View Full Product Details