Gene Transcripts

Gene Symbol
Pcp4
Entrez Gene ID
18546
Gene Name
Purkinje cell protein 4
Gene Aliases
P16Rimb19, Pcp-4, Pep19
Chromosome Location
Chr. 16: 96467606 - 96525789 on Build GRCm38
UniGene ID
Mm.5023
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008791.2 NP_032817.1 3 355
GenBank mRNA BC061164.1 - 3 355
X17320.1 - 3 194

View Full Product Details