Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016737.2 | NP_058017.1 | 2 | 233 | |
GenBank mRNA | AK075988.1 | - | 2 | 171 | |
AK088494.1 | - | 2 | 175 | ||
AK134171.1 | - | 2 | 149 | ||
AK149493.1 | - | 2 | 149 | ||
AK161645.1 | - | 2 | 152 | ||
AK167273.1 | - | 2 | 167 | ||
AK168181.1 | - | 2 | 149 | ||
AK181280.1 | - | 2 | 106 | ||
AK204291.1 | - | 2 | 106 | ||
BC003794.1 | - | 2 | 142 | 3590457 | |
CJ155828.1 | - | 2 | 234 | ||
U27830.1 | - | 2 | 150 |