Gene Transcripts

Gene Symbol
Ssr2
Entrez Gene ID
66256
Gene Name
signal sequence receptor, beta
Gene Aliases
1500032E05Rik, AI315033, AU020133, TLAP, TRAPB, TRAPbeta
Chromosome Location
Chr. 3: 88579584 - 88588420 on Build GRCm38
UniGene ID
Mm.486239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025448.3 NP_079724.1 3, 4 565
GenBank mRNA AK005340.1 - 4, 5 436
AK007538.1 - 1, 2 32
AK007720.1 - 1, 2 32
AK010565.1 - 4, 5 403
AK012281.1 - 3, 4 393
AK020145.1 - 4, 5 386
AK050505.1 - 3, 4 566
AK084530.1 - 4, 5 403
AK213548.1 - 2, 3 113
AK214352.1 - 1, 2 113
BC010214.1 - 3, 4 549 3497894

View Full Product Details