Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032394.3 | NP_115770.2 | 41 | 5647 | |
GenBank mRNA | AF242411.1 | - | 41 | 5684 | |
AK220152.1 | - | 16 | 2228 | ||
BC026416.1 | - | 14 | 1991 | ||
BC138338.1 | - | 41 | 5647 | 8861358 | |
BC138341.1 | - | 41 | 5647 | 8861361 |