Gene Transcripts

Gene Symbol
Grin3b
Entrez Gene ID
170483
Gene Name
glutamate receptor, ionotropic, NMDA3B
Gene Aliases
GluN3B, NR3B
Chromosome Location
Chr. 10: 79970397 - 79977190 on Build GRCm38
UniGene ID
Mm.391566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130455.2 NP_569722.1 2 775
GenBank mRNA AF396649.1 - 2 784
AY062435.1 - 2 775

View Full Product Details