Gene Transcripts
- Gene Symbol
- Vmn1r73
- Entrez Gene ID
- 171237
- Gene Name
- vomeronasal 1 receptor 73
- Gene Aliases
- V1rg2
- Chromosome Location
- Chr. 7: 11756257 - 11757168 on Build GRCm38
- UniGene ID
- Mm.377193
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134203.1 |
NP_598964.1
|
1 |
355 |
|
View Full Product Details