Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CESA2
Entrez Gene ID
830090
Gene Name
cellulose synthase A2
Gene Aliases
AT4G39350, ATCESA2, ATH-A, CELLULOSE SYNTHASE, T22F8.250, T22F8_250, cellulose synthase A2
Chromosome Location
Chr.4: 18296388 - 18302192 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120095.4 NP_195645.1 5 - 6 1712 114
GenBank mRNA AF027173.2 - 5 - 6 1192 114
KJ139038.1 - 5 - 6 1022 114

View Full Product Details