Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT2G18720
Entrez Gene ID
816387
Gene Name
Translation elongation factor EF1A/initiation factor IF2gamma family protein
Gene Aliases
AT2G18720, MSF3.10, MSF3_10
Chromosome Location
Chr.2: 8114785 - 8117085 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335603.1 NP_001324883.1 5 - 6 742 75
NM_001335604.1 NP_001324884.1 2 - 3 377 75
NM_127428.2 NP_179462.2 4 - 5 676 75

View Full Product Details