Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAA8
Entrez Gene ID
816798
Gene Name
indoleacetic acid-induced protein 8
Gene Aliases
AT2G22670, T9I22.11, T9I22_11, indoleacetic acid-induced protein 8
Chromosome Location
Chr.2: 9636346 - 9638761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084474.1 NP_001077943.1 2 - 3 668 117
NM_001202650.1 NP_001189579.1 3 - 4 867 117
NM_001335809.1 NP_001318267.1 2 - 3 1002 117
NM_001335810.1 NP_001324309.1 2 - 3 1015 117
NM_127832.3 NP_179852.1 2 - 3 811 117
GenBank mRNA AF334714.1 - 1 - 2 489 117
AY059905.1 - 2 - 3 667 117
AY070399.1 - 2 - 3 665 117
AY087635.1 - 2 - 3 718 117
AY096452.1 - 1 - 2 489 117
AY114671.1 - 1 - 2 489 117
U18410.1 - 2 - 3 629 117

View Full Product Details