Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P5CS2
Entrez Gene ID
824727
Gene Name
delta 1-pyrroline-5-carboxylate synthase 2
Gene Aliases
AT3G55610, delta 1-pyrroline-5-carboxylate synthase 2
Chromosome Location
Chr.3: 20624020 - 20629295 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125374.1 NP_001118846.1 9 - 10 1277 117
NM_115419.5 NP_191120.2 9 - 10 1479 117
GenBank mRNA AY091766.1 - 9 - 10 1277 117
Y09355.1 - - 1247 117

View Full Product Details