Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SDH3-1
- Entrez Gene ID
-
830819
- Gene Name
- succinate dehydrogenase 3-1
- Gene Aliases
-
AT5G09600, F17I14.210, F17I14_210, succinate dehydrogenase 3-1
- Chromosome Location
-
Chr.5: 2978825 - 2981096 on Build TAIR10.1
- Gene Symbol
- SDH3-2
- Entrez Gene ID
-
3770570
- Gene Name
- succinate dehydrogenase 3-2
- Gene Aliases
-
AT4G32210, F10M6.150, F10M6_150, succinate dehydrogenase 3-2
- Chromosome Location
-
Chr.4: 15556397 - 15558167 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119373.3 |
NP_194948.2 |
1 - 2 |
203 |
55 |
View Full Product Details