Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDH3-1
Entrez Gene ID
830819
Gene Name
succinate dehydrogenase 3-1
Gene Aliases
AT5G09600, F17I14.210, F17I14_210, succinate dehydrogenase 3-1
Chromosome Location
Chr.5: 2978825 - 2981096 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203339.2 NP_001190268.1 3 - 4 431 116
NM_120997.4 NP_196522.1 2 - 3 260 116
GenBank mRNA AY070732.1 - 2 - 3 231 116
AY074861.1 - 2 - 3 211 116
BT010154.1 - 2 - 3 147 116

Gene Symbol
SDH3-2
Entrez Gene ID
3770570
Gene Name
succinate dehydrogenase 3-2
Gene Aliases
AT4G32210, F10M6.150, F10M6_150, succinate dehydrogenase 3-2
Chromosome Location
Chr.4: 15556397 - 15558167 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119373.3 NP_194948.2 2 - 3 273 116

View Full Product Details