Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BON1
Entrez Gene ID
836311
Gene Name
Calcium-dependent phospholipid-binding Copine family protein
Gene Aliases
AT5G61900, BON, BONZAI 1, COPINE 1, CPN1, K22G18.2, K22G18_2
Chromosome Location
Chr.5: 24855661 - 24859341 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125583.4 NP_568944.1 2 - 3 238 66
NM_203248.2 NP_974977.1 1 - 2 273 66
GenBank mRNA AX380787.1 - 1 - 2 195 66
AY062824.1 - 2 - 3 229 66
BT010555.1 - 1 - 2 151 66

Gene Symbol
AT5G61910
Entrez Gene ID
836312
Gene Name
DCD (Development and Cell Death) domain protein
Gene Aliases
AT5G61910, K22G18.3, K22G18_3
Chromosome Location
Chr.5: 24855621 - 24864172 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203661.1 NP_001190590.1 8 - 9 2674 66

View Full Product Details