Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFC
Entrez Gene ID
831171
Gene Name
ARF GTPase-activating protein
Gene Aliases
AGD3, ARF-GAP DOMAIN3, ASCULAR NETWORK DEFECTIVE 3, AT5G13300, SCARFACE, T31B5.120, T31B5_120, VAN3
Chromosome Location
Chr.5: 4255547 - 4262568 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121333.4 NP_196834.3 4 - 5 843 97
GenBank mRNA AB194395.1 - 4 - 5 293 97

View Full Product Details