Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATHB13
Entrez Gene ID
843314
Gene Name
Homeobox-leucine zipper protein family
Gene Aliases
AT1G69780, HOMEODOMAIN LEUCINE-ZIPPER PROTEIN ATHB13, T6C23.2, T6C23_2
Chromosome Location
Chr.1: 26258788 - 26260781 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105646.3 NP_177136.1 2 - 3 898 151
GenBank mRNA AF208044.1 - 2 - 3 719 151
AY057572.1 - 2 - 3 744 151
AY133645.1 - 2 - 3 520 151

View Full Product Details