Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAA1
Entrez Gene ID
843393
Gene Name
tryptophan aminotransferase of Arabidopsis 1
Gene Aliases
AT1G70560, CKRC1, F24J13.13, F24J13_13, SAV3, SHADE AVOIDANCE 3, WEAK ETHYLENE INSENSITIVE 8, WEI8, cytokinin induced root curling 1, tryptophan aminotransferase of Arabidopsis 1
Chromosome Location
Chr.1: 26604855 - 26607652 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105724.3 NP_177213.1 2 - 3 432 99
GenBank mRNA AK117208.1 - 2 - 3 432 99
BT005339.1 - 2 - 3 393 99

View Full Product Details