Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHR11
Entrez Gene ID
819814
Gene Name
chromatin-remodeling protein 11
Gene Aliases
AT3G06400, F24P17.13, F24P17_13, chromatin-remodeling protein 11
Chromosome Location
Chr.3: 1940907 - 1946980 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202897.1 NP_001189826.1 19 - 20 2559 131
NM_111515.5 NP_187291.2 19 - 20 2619 131

View Full Product Details