Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VIIIB
Entrez Gene ID
828845
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT4G27370, ATVIIIB, F27G19.14, MYOSIN VIII B
Chromosome Location
Chr.4: 13694689 - 13701119 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341848.1 NP_001328308.1 22 - 23 2931 150
NM_001341849.1 NP_001328309.1 21 - 22 2870 150
NM_001341850.1 NP_001328307.1 21 - 22 2947 150
NM_118871.6 NP_194467.5 20 - 21 2738 150

View Full Product Details