Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MSL5
- Entrez Gene ID
-
820710
- Gene Name
- mechanosensitive channel of small conductance-like 5
- Gene Aliases
-
AT3G14810, mechanosensitive channel of small conductance-like 5
- Chromosome Location
-
Chr.3: 4970944 - 4974534 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112342.4 |
NP_188099.2 |
2 - 3 |
2146 |
80 |
View Full Product Details