Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIE
Entrez Gene ID
821622
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT3G20740, FERTILIZATION-INDEPENDENT ENDOSPERM, FERTILIZATION-INDEPENDENT ENDOSPERM 1, FIE1, FIS3
Chromosome Location
Chr.3: 7248809 - 7252452 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112965.1 NP_188710.1 5 - 6 732 77
GenBank mRNA AF129516.1 - 5 - 6 732 77
AK117114.1 - 5 - 6 589 77
BT005326.1 - 5 - 6 534 77

View Full Product Details