Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSH5
Entrez Gene ID
28721277
Gene Name
LIGHT-DEPENDENT SHORT HYPOCOTYLS-like protein (DUF640)
Gene Aliases
AT5G58500, LIGHT SENSITIVE HYPOCOTYLS 5, MQJ2.11, MQJ2_11
Chromosome Location
Chr.5: 23645062 - 23646347 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345303.1 NP_001318830.1 1 - 1 727 72
GenBank mRNA BT010401.1 - 1 - 1 463 72

View Full Product Details