Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIPS3
Entrez Gene ID
830881
Gene Name
myo-inositol-1-phosphate synthase 3
Gene Aliases
AT5G10170, ATMIPS3, MYO-INOSITOL-1-PHOSTPATE SYNTHASE 3, T31P16.160, T31P16_160, myo-inositol-1-phosphate synthase 3
Chromosome Location
Chr.5: 3187277 - 3190321 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121055.4 NP_196579.1 1 - 2 350 84
GenBank mRNA AY065415.1 - 1 - 2 280 84
AY096554.1 - 1 - 2 190 84

View Full Product Details