Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP85
Entrez Gene ID
821835
Gene Name
cupin family protein
Gene Aliases
AT3G22640
Chromosome Location
Chr.3: 8011724 - 8013941 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113163.4 NP_566714.1 1 - 2 620 71
GenBank mRNA AY058085.1 - 1 - 2 581 71
AY090307.1 - 1 - 2 562 71

View Full Product Details