Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHY1
Entrez Gene ID
836724
Gene Name
beta-hydroxyisobutyryl-CoA hydrolase 1
Gene Aliases
AT5G65940, COA-THIOESTER HYDROLASE, K14B20.11, K14B20_11, beta-hydroxyisobutyryl-CoA hydrolase 1
Chromosome Location
Chr.5: 26376578 - 26379681 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037078.2 NP_001032155.1 5 - 6 405 100
NM_001085335.1 NP_001078804.1 5 - 6 405 100
NM_001345728.1 NP_001330538.1 5 - 6 430 100
NM_125991.4 NP_201395.1 5 - 6 405 100
GenBank mRNA AF276301.1 - 5 - 6 359 100
AK316713.1 - 5 - 6 406 100
BT000956.1 - 5 - 6 331 100

View Full Product Details