Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATERDJ2A
Entrez Gene ID
844334
Gene Name
DnaJ / Sec63 Brl domains-containing protein
Gene Aliases
AT1G79940, F19K16.10, F19K16_10
Chromosome Location
Chr.1: 30069519 - 30073440 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036229.3 NP_001031306.2 7 - 8 1091 63
NM_001124151.1 NP_001117623.1 7 - 8 1082 63
NM_001198516.1 NP_001185445.1 7 - 8 1074 63
NM_106643.5 NP_178112.2 7 - 8 1063 63
GenBank mRNA AK227714.1 - 7 - 8 1009 63

View Full Product Details