Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DWF4
Entrez Gene ID
824229
Gene Name
Cytochrome P450 superfamily protein
Gene Aliases
AT3G50660, AtDWF4, CLM, CLOMAZONE-RESISTANT, CYP90B1, CYTOCHROME P450 90B1, DWARF 4, PARTIALLY SUPPRESSING COI1 INSENSITIVITY TO JA 1, PARTIALLY SUPPRESSING COI1 INSENSITIVITY TO JA 1, PSC1, SAV1, SHADE AVOIDANCE 1, SNP2, STEROID 22-ALPHA-HYDROXYLASE, SUPPRESSOR OF NPH4 2
Chromosome Location
Chr.3: 18813949 - 18817405 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339479.1 NP_001327730.1 6 - 7 1418 65
NM_114926.4 NP_190635.1 6 - 7 1418 65
GenBank mRNA AF412114.1 - 6 - 7 1209 65
AY090266.1 - 6 - 7 1181 65

View Full Product Details