Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ELF5A-3
- Entrez Gene ID
-
843273
- Gene Name
- eukaryotic elongation factor 5A-3
- Gene Aliases
-
AT1G69410, ATELF5A-3, EUKARYOTIC ELONGATION FACTOR 5A-3, F10D13.8, F10D13_8, eukaryotic elongation factor 5A-3
- Chromosome Location
-
Chr.1: 26089030 - 26090734 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105608.4 |
NP_177100.1 |
3 - 4 |
568 |
146 |
GenBank mRNA |
AY087040.1 |
- |
3 - 4 |
393 |
146 |
View Full Product Details