Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSM12
Entrez Gene ID
538005
Gene Name
LSM12 homolog
Gene Aliases
-
Chromosome Location
Chr.19: 43884990 - 43904700 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076098.2 NP_001069566.2 3 - 4 461 128
GenBank mRNA BC120190.1 - 3 - 4 461 128
DT830787.1 - 3 - 4 435 128
EE246344.1 - 3 - 4 461 128

View Full Product Details