Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTA1
Entrez Gene ID
281592
Gene Name
actin alpha 1, skeletal muscle
Gene Aliases
-
Chromosome Location
Chr.28: 1400443 - 1403202 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174225.1 NP_776650.1 1 - 2 97 60
GenBank mRNA BC134665.1 - 1 - 2 89 60

View Full Product Details