Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANXA8L1
Entrez Gene ID
281627
Gene Name
annexin A8-like 1
Gene Aliases
ANXA8
Chromosome Location
Chr.28: 41953896 - 41970251 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174241.2 NP_776666.1 7 - 8 601 85
GenBank mRNA AF417637.1 - 7 - 8 601 85
BC113321.1 - 7 - 8 650 85
BT021645.1 - 7 - 8 633 85
BT021646.1 - 7 - 8 633 85

View Full Product Details