Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD1
Entrez Gene ID
510774
Gene Name
abhydrolase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 72508608 - 72515678 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035094.2 NP_001030266.1 1 - 2 150 69
GenBank mRNA BC102483.1 - 1 - 2 152 69

View Full Product Details