Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLBA1
Entrez Gene ID
512628
Gene Name
clathrin binding box of aftiphilin containing 1
Gene Aliases
C14orf79, C21H14orf79
Chromosome Location
Chr.21: 69403996 - 69409670 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075573.1 NP_001069041.1 2 - 3 800 70
GenBank mRNA BC118243.1 - 2 - 3 800 70

View Full Product Details