Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JOSD1
Entrez Gene ID
510781
Gene Name
Josephin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 110317480 - 110329100 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014898.2 NP_001014898.1 1 - 2 800 77
GenBank mRNA BC140477.1 - 1 - 2 800 77
BT020624.1 - 1 - 2 514 77

View Full Product Details