Product Details

SNP ID
rs75419063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10397739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATAAATAAATATATACAGAGA[C/T]GTGAACACTGATTCCCTTATATAAC
Phenotype
MIM: 107580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFAP2A PubMed Links

Gene Details

Gene
TFAP2A
Gene Name
transcription factor AP-2 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032280.2 2220 UTR 3 NP_001027451.1
NM_001042425.1 2220 UTR 3 NP_001035890.1
NM_003220.2 2220 UTR 3 NP_003211.1
XM_006715175.2 2220 UTR 3 XP_006715238.1
XM_011514833.2 2220 UTR 3 XP_011513135.1
XM_017011232.1 2220 UTR 3 XP_016866721.1

View Full Product Details