Product Details

SNP ID
rs79870417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34993232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACCAGTGCTGGTCCAGTGGCTG[G/T]GATGGGAAAAGATTATTACAAGATT
Phenotype
MIM: 611328
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB5 PubMed Links

Gene Details

Gene
DNAJB5
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135004.2 143 Missense Mutation GGG,GTG G114V NP_001128476.2
NM_001135005.2 143 Missense Mutation GGG,GTG G72V NP_001128477.1
NM_012266.5 143 UTR 5 NP_036398.3
XM_005251428.3 143 UTR 5 XP_005251485.1
XM_006716751.2 143 UTR 5 XP_006716814.1
XM_011517842.2 143 Missense Mutation GGG,GTG G72V XP_011516144.1
Gene
DNAJB5-AS1
Gene Name
DNAJB5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details