Product Details

SNP ID
rs75439362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133310190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTGCTTGGTAGAAGGCTGCAC[A/T]GAGAAGTTCAAGACCAGCAGAGACC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TUBGCP2 PubMed Links

Gene Details

Gene
TUBGCP2
Gene Name
tubulin gamma complex associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256617.1 511 Intron NP_001243546.1
NM_001256618.1 511 Intron NP_001243547.1
NM_006659.3 511 Intron NP_006650.1
XM_017015595.1 511 Intron XP_016871084.1
XM_017015596.1 511 Intron XP_016871085.1
Gene
ZNF511
Gene Name
zinc finger protein 511
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145806.3 511 Silent Mutation ACA,ACT T152T NP_665805.2

View Full Product Details