Product Details

SNP ID
rs74927124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1936083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTCGTGGGTGAGCCTCATTT[A/G]TTCTTCTCTGCGTCCTGCTTTCTCC
Phenotype
MIM: 610461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTN4RL1 PubMed Links

Gene Details

Gene
RTN4RL1
Gene Name
reticulon 4 receptor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178568.3 2219 UTR 3 NP_848663.1

View Full Product Details