Product Details

SNP ID
rs75008465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2482369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAAATTGAGGATTCACTGGGGA[G/T]TGAGGGCAGGACTGGGCAGAGGAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF343 PubMed Links

Gene Details

Gene
ZNF343
Gene Name
zinc finger protein 343
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282495.1 2944 UTR 3 NP_001269424.1
NM_001282496.1 2944 UTR 3 NP_001269425.1
NM_001282497.1 2944 UTR 3 NP_001269426.1
NM_001282498.1 2944 UTR 3 NP_001269427.1
NM_001282499.1 2944 Intron NP_001269428.1
NM_001321800.1 2944 UTR 3 NP_001308729.1
NM_001321801.1 2944 UTR 3 NP_001308730.1
NM_001321802.1 2944 UTR 3 NP_001308731.1
NM_001321803.1 2944 UTR 3 NP_001308732.1
NM_001321805.1 2944 UTR 3 NP_001308734.1
NM_024325.5 2944 UTR 3 NP_077301.4
XM_017028062.1 2944 UTR 3 XP_016883551.1

View Full Product Details