Product Details

SNP ID
rs76357418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26663337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTTGGAGGCATCTGTCCACCCA[C/T]GTGGTTCCAGACACGTTCATGTGGC
Phenotype
MIM: 611082
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIAT PubMed Links

Gene Details

Gene
MIAT
Gene Name
myocardial infarction associated transcript (non-protein coding)
There are no transcripts associated with this gene.

Gene
MIATNB
Gene Name
MIAT neighbor (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details