Product Details
- SNP ID
-
rs74473757
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:45006333 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCTGTGCAGGGACGGCAGCCCA[A/G]GGGGCGGCGGGCTGAGGAGCAGTGC
- Phenotype
-
MIM: 604994
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SIX2
PubMed Links
Gene Details
- Gene
- SIX2
- Gene Name
- SIX homeobox 2
View Full Product Details