Product Details

SNP ID
rs78519734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:31545076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCTTTTATTTTAATATGCTAA[A/G]CAGCTTTATGGCATTTGAAGGAACA
Phenotype
MIM: 608605
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STT3B PubMed Links

Gene Details

Gene
STT3B
Gene Name
STT3B, catalytic subunit of the oligosaccharyltransferase complex
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178862.2 Intron NP_849193.1
XM_011533465.1 Intron XP_011531767.1
XM_017005857.1 Intron XP_016861346.1
XM_017005858.1 Intron XP_016861347.1
XM_017005859.1 Intron XP_016861348.1

View Full Product Details