Product Details

SNP ID
rs76859970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48404164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTGGCCAAGGCCAGTGTTCAG[A/G]AACAGTACAGTCTCCCCAGGGGCCT
Phenotype
MIM: 609075 MIM: 601053
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXW12 PubMed Links

Gene Details

Gene
FBXW12
Gene Name
F-box and WD repeat domain containing 12
There are no transcripts associated with this gene.

Gene
PLXNB1
Gene Name
plexin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130082.2 10418 UTR 3 NP_001123554.1
NM_002673.5 10418 UTR 3 NP_002664.2
XM_011533833.2 10418 UTR 3 XP_011532135.1
XM_011533834.1 10418 UTR 3 XP_011532136.1
XM_011533835.1 10418 UTR 3 XP_011532137.1
XM_011533836.1 10418 UTR 3 XP_011532138.1
XM_011533837.2 10418 UTR 3 XP_011532139.1
XM_017006630.1 10418 UTR 3 XP_016862119.1
XM_017006631.1 10418 UTR 3 XP_016862120.1

View Full Product Details