Product Details

SNP ID
rs78700237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50607838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTCCTTAGGCATAGGCAGGGC[C/T]GGGGTGGGAGCAGGATCGGGGCTGT
Phenotype
MIM: 602441 MIM: 602130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CISH PubMed Links

Gene Details

Gene
CISH
Gene Name
cytokine inducible SH2 containing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013324.5 3549 Silent Mutation NP_037456.5
NM_145071.2 3549 Silent Mutation NP_659508.1
XM_011533329.2 3549 Silent Mutation XP_011531631.1
Gene
MAPKAPK3
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
There are no transcripts associated with this gene.

View Full Product Details