Product Details

SNP ID
rs78191714
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136429840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAGGGGGCGTTAGGAGGGCAC[C/T]CAGGGCCAGGAGGAGGGGGCGTTAG
Phenotype
MIM: 613037 MIM: 613036
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
INPP5E PubMed Links

Gene Details

Gene
INPP5E
Gene Name
inositol polyphosphate-5-phosphatase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318502.1 Intron NP_001305431.1
NM_019892.5 Intron NP_063945.2
XM_017014926.1 Intron XP_016870415.1
Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
There are no transcripts associated with this gene.

View Full Product Details