Product Details

SNP ID
rs74822962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137252074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAGGTACCAGGTGCCGAGCCC[A/G]TCCGTACGAGAGTCCTCCCCACACC
Phenotype
MIM: 611180 MIM: 602660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf173-AS1 PubMed Links

Gene Details

Gene
C9orf173-AS1
Gene Name
C9orf173 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM166A
Gene Name
family with sequence similarity 166 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001710.2 368 Intron NP_001001710.1
XM_006717108.2 368 Intron XP_006717171.2
XM_011518694.1 368 Intron XP_011516996.1
XM_011518695.1 368 Intron XP_011516997.1
XM_011518696.2 368 Intron XP_011516998.1
XM_011518697.1 368 Intron XP_011516999.1
XM_017014718.1 368 Intron XP_016870207.1
Gene
NELFB
Gene Name
negative elongation factor complex member B
There are no transcripts associated with this gene.

Gene
STPG3
Gene Name
sperm-tail PG-rich repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004353.3 368 Silent Mutation CCA,CCG P114P NP_001004353.2
NM_001256699.1 368 Silent Mutation CCA,CCG P114P NP_001243628.1
NM_001256700.1 368 Silent Mutation CCA,CCG P114P NP_001243629.1
NM_001256701.1 368 Silent Mutation CCA,CCG P114P NP_001243630.1
XM_006717113.2 368 Silent Mutation CCA,CCG P114P XP_006717176.1
XM_006717115.2 368 Silent Mutation CCA,CCG P114P XP_006717178.1
Gene
TUBB4B
Gene Name
tubulin beta 4B class IVb
There are no transcripts associated with this gene.

View Full Product Details