Product Details

SNP ID
rs76692998
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:65149837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGTTTTCTGTTGTGGGTATTG[C/T]TACCTCAAGTGCGCCTATTTTGATT
Phenotype
MIM: 103030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AK4 PubMed Links

Gene Details

Gene
AK4
Gene Name
adenylate kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005353.2 Intron NP_001005353.1
NM_013410.3 Intron NP_037542.1
NM_203464.2 Intron NP_982289.1
XM_017000612.1 Intron XP_016856101.1
XM_017000613.1 Intron XP_016856102.1

View Full Product Details